Learn More
Cytoskeleton, Inc. TUBULIN POLYMERIZ ASSAY KIT
Supplier:  Cytoskeleton, Inc. BK004P
Encompass
Tubulin polymerization HTS assay using > 97% pure tubulin, OD based - Porcine (BK004P)
Catalog No. NC9969876