Learn More
Selleck Chemical LLC Tucatinib
SDP
Supplier:  Selleck Chemical LLC S8362100MG
Encompass_Preferred
Tucatinib (Irbinitinib ONT-380 ARRY-380) is an oral potent selective reversible and ATP-competitive small-molecule inhibitor of ErbB-2 (also called HER2) with IC50s of 8 nM and 7 nM for ErbB-2 and p95 HER2 respectively in cell-based assays showing 500-fold selective for HER2 vs EGFR It has potential antineoplastic activity
Catalog No. 50-000-00047