Learn More
Accuflow Systems TuFlux SIL 1/4"x3/8" (1/4 x 3/8 inch, 6,4 x 9,6 mm)
Supplier:  Accuflow Systems FSA117426
Encompass_Preferred
Manufactured by Sartorius, FSA117426 Tuflux SIL ¼’’ x ⅜’’, 6.4 x 9.5 mm wall thickness 1.6 mm, 1 coil = 50 m Piece $260 1 $260 1 piece
Catalog No. NC2187007