Learn More
Selleck Chemical LLC Tulobuterol E7406-50MG
SDP
Supplier:  Selleck Chemical LLC E740650MG
Encompass_Preferred
Tulobuterol (C-78 free base) is a long-acting 2-adrenoceptor agonist which reduces the frequency of exacerbations of chronic obstructive pulmonary disease and bronchial asthma Tulobuterol is also a sympathomimetic agent used as a transdermal patch and increases normal diaphragm muscle strength
Catalog No. 50-000-42007