Learn More
Creative Biomart Tumor Necrosis Factor alpha (TNF-alpha)
Supplier:  Creative Biomart THP0220
Encompass_Preferred
TNF-alpha protein (Val 77-Leu 233) is produced mainly by macrophages, and large amounts of this cytokine are released in response to lipopolysaccharide, other bacterial products, and Interleukin-1 (IL-1). TNF-alpha is involved in fighting against the tumorigenesis, thus, is regarded as a molecular insight in cancer treatment.
Catalog No. 50-208-4271