Learn More
Apexbio Technology LLC tumor protein p53 binding protein fragment [Homo sapiens]/[Mus musculus] 5mg
SDP
Supplier:  Apexbio Technology LLC A10945
Encompass_Preferred
tumor protein p53 binding protein fragment is a protein fragment derived from Homo sapiens and Mus musculus that interacts with the central DNA-binding domain of tumor protein p53 It is designed to modulate p53-dependent transcriptional activation thereby influencing DNA damage response pathways tumor protein p53 binding protein fragment exerts its biological activity by participating in the cellular DNA damage response network particularly through ATM-dependent phosphorylation and localization to DNA damage foci following double-strand breaks Based on these properties tumor protein p53 binding protein fragment holds research potential in studying DNA damage signaling genomic stability and tumorigenesis mechanisms
Catalog No. 50-000-96613