Learn More
World Precision Instrument Replacement Lamp for SurgioScope
SDP
Supplier:  World Precision Instrument 500162
Encompass
WPI’s improved precision SurgioScope (now with five magnification steps) is a portable, high quality surgical microscope offering outstanding image quality and value. Incorporating an agile extension arm and excellent working distance objectives, the SurgioScope provides convenient movement and maneuverability necessary for accurate positioning. These optional accessories are available for the SurgioScope Surgical Microscope. Chose from a 0.5" CS-Mount Adapter, a Beam Splitter, four different objectives, or a replacement light bulb. The CS-Mount Adaptor lets you attach a camera to your microscope. This also requires the Beam Splitter to allow you to attach the CS-Mount Adapter and camera. Available objectives include the F100, F250, F300, and F350. And the replacement lamp is a 12V, 100W light bulb suitable for the SurgioScope Surgical Microscope.
Catalog No. NC1077412