Learn More
Perkin Elmer US LLC UV/Vis Detector Tungsten Lamp for use with Series 200/785, ABD 7XX Series, ABI 759A, ABI 783 (A), and ABI 980.
Supplier:  Perkin Elmer US LLC N2920146
Encompass_Preferred
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer tungsten source provides outstanding ultraviolet and true visible performance. Exceptional performance anywhere in the detector's 190-700 nm wavelength range Lamp changes are quick and easy due toa unique self-aligning lamp mount
Catalog No. 50-904-4155