Learn More
Fine Science Tools Fine Scissors, Tungsten Carbide & ToughCut, 9 cm
SDP
Supplier:  Fine Science Tools 1455809
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Tungsten Carbide/ Length: 9cm/ Serrated: Yes/ Feature: Tungsten Carbide & ToughCut
Catalog No. NC9859721