Learn More
Grainger Turning Insert Diamond VBMT 221HQ PV72
Supplier:  Grainger 184ZR4
Encompass_Preferred
The Grainger Approved Turning Insert Diamond is a turning insert made of diamond for use on ferrous metals. It has a .031" thickness, .250" width, and .250" length. The insert has a .125" hole size and is uncoated. It has an ISO classification of DCMT070204 and features a diamond grade of MD100. The turning insert is designed for high speed machining applications where an ultra-sharp cutting edge is required. It provides excellent wear resistance and can machine hardened steels up to 65 HRC. The diamond insert enables smooth cutting action and superior surface finishes.
Catalog No. 50-276-3822