Learn More
Grainger Turning Insert Diamond VNMG 3305PP PV7
Supplier:  Grainger 184ZU4
Encompass_Preferred
The Grainger Approved Turning Insert is a carbide insert designed for turning operations. It features a CNMG 432 geometry with a TiAlN PVD coating for wear resistance. The insert has a hole for screw mounting and is made of micro-grain carbide for strength and durability. It can be used for continuous and interrupted cuts in steel, stainless steel, cast iron, and non-ferrous metals. The turning insert measures 0.4" x 0.4" x 0.2" thick and is sold individually. It is manufactured in the USA by Grainger Industrial Supply.
Catalog No. 50-276-4236