Learn More
Grainger Turning Insert Diamond VNMG 331PP TN62
Supplier:  Grainger 184ZU6
Encompass_Preferred
The Kyocera VNMG331PQTN62 turning insert is a diamond-tipped cutting tool for metalworking. It features a VNMG331 geometry with a .031" nose radius and .062" thickness. The insert is made of TN62 micrograin carbide for high abrasion resistance and toughness. It has a PVD TiAlN coating for heat and wear resistance. The insert is designed for high speed machining in steel, cast iron, and other materials. It provides precise cutting edges and smooth surface finishes. The VNMG331PQTN62 is optimal for finishing operations like profiling, facing, and shoulder milling.
Catalog No. 50-276-4338