Learn More
Grainger Turning Insert Trigon WNMG 433WE PV720
Supplier:  Grainger 185GM3
Encompass_Preferred
The Grainger 185GM3 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase open drip proof motor with a 48Y frame and features permanent split capacitor for high starting torque. The motor has a 0.8 service factor and is thermally protected. It can be used for driving fans, blowers, pumps and other light industrial applications. The motor dimensions are 5-7/8" diameter x 7-3/8" long and it weighs 12 pounds.
Catalog No. 50-276-4180