Learn More
Medchemexpress LLC Tuspetinib | 2294874-49-8 | 99.9% | C29H33ClN6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14501510MG
Encompass_Preferred
Tuspetinib is a selective, orally active FLT3 inhibitor supplied as a research-grade small molecule. It is available in solid form and as alternative salt/hydrate variants, with documented potency against FLT3 and characterized physicochemical data. For laboratory research use only; not for human therapeutic use.
  • Selective FLT3 inhibitor with low nanomolar potency.
  • Provided as free base, hydrate, and dihydrochloride forms.
  • High reported purity (~99.85%).
  • White to off-white solid suitable for analytical and biological assays.
  • Documented identifiers and data to support reproducible research.
Catalog No. 50-002-61403