Learn More
Selleck Chemical LLC Tuxobertinib S9786-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S978610MM/1ML
Encompass_Preferred
Tuxobertinib (BDTX-189) is a potent and selective inhibitor of allosteric EGFR and HER2 oncogenic mutations with Kd of 0 2 nM 0 76 nM 13 nM and 1 2 nM for EGFR HER2 BLK and RIPK2 reapectively BDTX-189 exhibits anticancer activity
Catalog No. 50-000-24406