Learn More
TARGETMOL CHEMICALS INC TV 3279 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T773325MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TV 3279 is a novel ChE-MAI inhibitor and the neuroprotective properties depend on their ability to induce the anti-apoptotic proteins PKC Bcl-2 Bcl-x and SOD and to block the nuclear translocation of the pro-apoptotic enzyme glyceraldehyde phosphate dehydrogenase in PC-12 and neuroblastoma cells. purity: 98%
Catalog No. 50-257-9156