Learn More
Selleck Chemical LLC TVB-3166-5mg
SDP
Supplier:  Selleck Chemical LLC S35765MG
Encompass_Preferred
TVB-3166 is an orally-available, reversible, potent and selective inhibitor of fatty acid synthase (FASN) with IC50 of 0.042 μM in an in vitro biochemical assay. TVB-3166 induces apoptosis, and inhibits in-vivo xenograft tumor growth.
Catalog No. 50-238-9608