Learn More
Medchemexpress LLC HY-120062 5mg Medchemexpress, TVB-3664 CAS:2097262-58-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1200625MG
Encompass_Preferred
Medchemexpress, HY-120062 5mg TVB-3664 CAS:2097262-58-1 TVB-3664 is an orally available, reversible, potent, selective and highly bioavailable fatty acid synthase (FASN) inhibitor, with IC50 values of 18 nM and 12 nM for human and mouse cell palmitate synthesis, respectively. TVB-3664 significantly reduces tubulin palmitoylation and mRNA expression[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9865