Learn More
Selleck Chemical LLC TW-37 10mM/1mL 877877-35-5
SDP
Supplier:  Selleck Chemical LLC S112110MM/1ML
Encompass_Preferred
TW-37 is a novel nonpeptide inhibitor to recombinant Bcl-2, Bcl-xL and Mcl-1 with Ki of 0.29 ?M, 1.11 ?M and 0.26 ?M in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0937