Learn More
Sigma Aldrich Fine Chemicals Biosciences TWEEN 80 Solution, 9005-65-6, MFCD00082107, 10mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P819210ML
Encompass
TWEEN 80 Solution (10%) non-ionic detergent is used for selective protein extraction and isolation of nuclei from mammalian cell lines.
Catalog No. NC1617536