Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Superalloy CX Biological Tweezers Style 4, 110 mm OAL
SDP

Supplier:  Electron Microscopy Sciences 783254SA

Encompass_Preferred

EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 110 mm
OAL: extra fine
Tip width: 0.03 mm

Catalog No. 50-192-9478


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.