Learn More
World Precision Instrument Dumont Tweezers #5, 11cm, Straight, 0.1 x 0.06mm Tips
SDP
Supplier:  World Precision Instrument 500342
Encompass
Dumostar Material Information
Highly resistant to metal fatigue, has great elasticity, and is durable (Rockwell Hardness measurement of 62). Resistant to mineral and organic acids, salt. Autoclavable. More likely to break than to bend. The maximum temperature this material can withstand is 550° C.
Catalog No. 50-822-451