Learn More
Cole-Parmer Aven Tools Tweezers, stainless steel, straight, strong precision tips, 5"L
Supplier:  Cole-Parmer 0728701
Encompass_Preferred
Recommended for micro electronics and precision assembly, these finely crafted stainless steel tweezers feature hand finished points for accuracy and excellent tip hardness. Anti-glare finish reduces eye strain. Anti-magnetic, anti-acid, and acid resistant.
Catalog No. NC9039011