Learn More
Electron Microscopy Sciences EMS Tweezers Style 5 with Replaceable PVDF Tips, 130 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7832538
Encompass_Preferred
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Very fine tips.
Material: PVDF
OAL: 130 mm
Catalog No. 50-193-0852