Learn More
World Precision Instrument Tweezers 11cm (4.33") Long, 4.5mm Tip, Black Epoxy Coating With Patented EZ Lever
SDP
Supplier:  World Precision Instrument WPB315AB
Encompass
Cutting tweezers with a black epoxy coating are a specialized surgical instrument designed for precision cutting in medical procedures. The black epoxy coating enhances the forceps' durability by providing corrosion resistance. It also reduces the glare under surgical lights or a microscope. The thumb lever in the handle serves as a unique and ergonomic mechanism, allowing you to easily squeeze the sharp blades together for controlled cutting. This feature not only ensures precise cutting but also offers a firm and comfortable grip during procedures. The forceps' versatility makes them suitable for various medical specialties, contributing to tasks such as suturing and tissue trimming. The ergonomic design, combined with the safety features like the thumb lever, enhances user comfort and instrument control. These tweezers may be sterilized using an autoclave or a chemical sterilant.
Catalog No. 50-253-8274