Learn More
Electron Microscopy Sciences Tweezers for handling sensitive electronic component feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. ESD safe.
SDP
Supplier:  Electron Microscopy Sciences 7832536
Encompass_Preferred
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical.
Catalog No. NC3807395