Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style 5B, 110 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7814012
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Super-fine points, with 35 deg curved tips.
OAL: 110 mm
Catalog No. 50-237-87