Learn More
Electron Microscopy Sciences EMS Tweezers Style 1, Titanium, Fine, accurate, strong Tips
SDP
Supplier:  Electron Microscopy Sciences 7851101
Encompass_Preferred
The tweezers are the Swiss highest quality. High precision tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: Titanium
Tips: Fine, accurate, strong
OAL: 120 mm
Catalog No. 50-193-0564