Learn More
ABclonal Technology TWF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15307500UL
Encompass_Preferred
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
Catalog No. 50-001-08026