Learn More
Gerstel TWICESTER EVAL KIT- 6 TWISTERS
SDP
Supplier:  Gerstel 01692020300
Encompass
GERSTEL Twicester Evaluation Kit - Version 1Allows up to 4 Twisters to be used for sampling in a single 10/20 mL vial. Consists of:3 x PDMS Twister 10 x 0,5 mm3 x EG Silicone Twister3 x Twister holder for 10/20 ml vials 1 x Tweezers
Catalog No. NC2584159