Learn More
OriGene Twist1 (NM_011658) Mouse Recombinant Protein, 20 ug
SDP
Supplier:  OriGene TP527370
Encompass_Preferred
Purified recombinant protein of Mouse twist basic helix-loop-helix transcription factor 1 (Twist1), with C-terminal MYC/DDK tag, expressed in HEK293T cells, 20ug
Catalog No. NC3243075