Learn More
ABclonal Technology TWIST1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A25134100UL
Encompass_Preferred
This gene encodes a basic helix-loop-helix (bHLH) transcription factor that plays an important role in embryonic development The encoded protein forms both homodimers and heterodimers that bind to DNA E box sequences and regulate the transcription of genes involved in cranial suture closure during skull development This protein may also regulate neural tube closure limb development and brown fat metabolism This gene is hypermethylated and overexpressed in multiple human cancers and the encoded protein promotes tumor cell invasion and metastasis as well as metastatic recurrence Mutations in this gene cause Saethre-Chotzen syndrome in human patients which is characterized by craniosynostosis ptosis and hypertelorism
Catalog No. 50-000-10719