Learn More
Electron Microscopy Sciences Two Part Conductive Silver Paint, 14 g kit
SDP
Supplier:  Electron Microscopy Sciences 1264214
Encompass_Preferred
This electrically conductive silver epoxy is used for adhering samples as well as solderless connections such as bonding in electric design, prototype and repair work, circuit board repair, surface mount connections, static discharge, shielding and grounding. It is also ideal for the bonding of heat sensitive components. Its curing time is within ten minutes at 100 degree F (38 degree C), or at room temperature. Conductivity is 0.001 ohm/cm.
Catalog No. 50-980-315