Learn More
Electron Microscopy Sciences TWO-PIECE PLASTC CONTNRS 256DR
SDP
Supplier:  Electron Microscopy Sciences 7261753
Encompass_Preferred
The two Piece Plastic Containers have a double think wall and are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe, and recyclable.
Catalog No. 50-241-3087