Learn More
BURKLE INC Two-way valve, PP/PE, Ø 0.43-0.51 in. (11-13 mm), NW 0.31 in. (8 mm), red/white
Supplier:  BURKLE INC 86050120
Encompass_Preferred
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25892