Learn More
Andwin Scientific TY-RAP XTR-HITEMP 7IN 1000PK
SDP
Supplier:  Andwin Scientific TYHT25M1000
Encompass_Preferred
The 7.31 x 0.185-inch THOMAS & BETTS TYHT25M Cable Tie is constructed of high temperature nylon 4.6. This is made to avoid snags on uneven surfaces and easier to pull through bulkheads. This product provides the maximum strength and the right tightness every use. Due to its design of having no notches, the Ty-Rap tie gives an infinite number of locking positions while fastening the tie at the precise bundle diameter needed. It also has a unique friction surface that lessens slippage during installation. The cable ties can operate within the temperature range of -40 to 302F. This is sold in a pack of 1000.
Catalog No. NC9067138