Learn More
Kirby Risk Electrical Supply Cable Tie, Natural Polyamide (Nylon 6.6) 1000/pack
Supplier:  Kirby Risk Electrical Supply TY23MPK
Encompass_Preferred
Cable Tie, Natural Polyamide (Nylon 6.6) for Temperatures up to 85 Degrees Celsius (185 F) for Indoor Applications, UL/EN/CSA62275 Type 2/21 Rated for AH-2 Plenum, Length of 91.95mm (3.62 Inches), Width of 2.29mm (0.09 Inches), Thickness of 0.75mm (0.03 Inches), Tensile Strength Rating of 80 Newtons (18 Pounds), Military Specified (MIL-SPEC MS3367-4-9), Bulk Pack
Catalog No. NC9756882