Learn More
Saint Gobain Performance Plastics Tube Tygon 1/2 x 3/4 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ACF00038
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 1/2 in. Diameter Outer 3/4 in. Thickness 1/8 in. Overall Length 50 ft. 15 m Clear
Catalog No. 50-206-8963