Learn More
AVANTOR MASTERFLEX (VWR LLC) Vacuum Tubing, 3/8""ID x 7/8""OD, 10 ft/pack
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0641430
Encompass
Transparent Vacuum Tubing, 3/8"ID x 7/8"OD, 10 ft/pack. Formulation: Tygon E-3603 vacuumApplications: General lab, food and beverage, and biopharm. Vacuum service of 29.9¿ Hg (760 mm Hg) at 73¿F (23¿C).Characteristics: Non-DEHP, phthalate-free plasticizer. Flexible, biodegradable, crack-resistant, nontoxic, and nonoxidizing. Clear.Certifications: USP Class VI and NSF 51Temperature range: Brittle temperature is ¿51F (¿46¿C).Sterilize: Ethylene oxide or autoclave for 30 min at 250¿F (121¿C), 15 psi (1 bar).
Catalog No. NC0646086