Learn More
US Plastic Corporation 1/8" ID x 1/4" OD x 1/16" Wall Tygon 2375 Ultra Chemical Resistant Tubing
SDP
Supplier:  US Plastic Corporation 5754750FT/DUP
Encompass
This tubing offers an unequaled combination of chemical resistance, clarity, and flexibility. It is virtually unaffected by acids, bases, ketones, salts, and alcohols, enabling it to meet many applications' requirements, from filling battery acid to handling hazardous material. With an exceptionally smooth inner bore, the potential for contamination is reduced because the particulate buildup is inhibited.
Catalog No. NC1923072