Learn More
Accuflow Systems Saint-Gobain Thermoplastic Tubing, High-Purity, 3/4 in ID, 1 in OD
Supplier:  Accuflow Systems ACG00053
Encompass
Tygon 2475 tubing is a crystal-clear, plasticizer-free thermoplastic designed for medical, diagnostic, and laboratory fluid transfer where low extractables, chemical resistance, and biocompatibility are required. It minimizes contamination risk and offers low gas permeation compared with silicone, making it suitable for reagent delivery, analyzer waste lines, cell and tissue transport, and other clean fluid-handling applications.
  • High-purity, plasticizer-free thermoplastic formulation.
  • Clear, crystal transparency for visual inspection of flow and bubbles.
  • Resists aggressive acids, bases, and common solvents.
  • Meets USP Class VI biocompatibility standards.
  • ID/OD available up to 1 in ID and 1-3/8 in OD; common size 3/4 in ID, 1 in OD.
  • Wall thickness range from 1/16 in to 3/16 in.
  • Low brittleness point (reported -108°F / -78°C); full operating range not specified.
Catalog No. 50-236-5792