Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, 5/16 in ID, 1/16 in Wall
Supplier:  Accuflow Systems ACF00022
Encompass_Preferred
Tygon S3 E-3603 tubing from Saint-Gobain is a crystal-clear, phthalate-free thermoplastic elastomer tubing designed for laboratory and vacuum applications. The 5/16 in inner diameter, 7/16 in outer diameter, 1/16 in wall thickness size is commonly supplied on 50 ft coils and is suitable for peristaltic pumps, instrumentation connections, venting, and drain lines.
  • Thermoplastic elastomer material provides chemical resistance.
  • Dimensions: 5/16 in ID, 7/16 in OD, 1/16 in wall.
  • Packaged in 50 ft (15.24 m) coil for extended runs.
  • Phthalate-free/non-DEHP formulation for sensitive applications.
  • Resistant to flex-fatigue and abrasion for longer service life.
  • Glassy smooth inner bore reduces buildup and contamination.
  • Capable of many laboratory vacuum applications (manufacturer data varies by size).
Catalog No. 50-236-5831