Learn More
Medchemexpress LLC HY-101762 5mg Medchemexpress, TyK2-IN-2 CAS:2098466-94-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1017625MG
Encompass_Preferred
Medchemexpress, HY-101762 5mg TyK2-IN-2 CAS:2098466-94-3 TyK2-IN-2 is an inhibitor of TYK2 , used for treatment of inflammatory and autoimmune diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2206