Learn More
Selleck Chemical LLC Tyloxapol S4578-100mg
SDP
Supplier:  Selleck Chemical LLC S4578100MG
Encompass_Preferred
Tyloxapol (Triton WR1339) is a nonionic liquid polymer of the alkyl aryl polyether alcohol type used as a surfactant to aid liquefaction removal of mucopurulent and bronchopulmonary secretions It also blocks plasma lipolytic activity and thus the breakdown of triglyceride-rich lipoproteins Tyloxapol can be used to induce animal models of Hyperlipidemia Atherosclerosis
Catalog No. 50-311-0435