Learn More
Medchemexpress LLC Tyloxapol | 25301-02-4 | 10 G
SDP
Supplier:  Medchemexpress LLC HYB106810G
Encompass_Preferred
Tyloxapol is a nonionic liquid polymer of the alkyl aryl polyether alcohol type, functioning as a surface active stabilizer. It is utilized to induce hyperlipidemia in animals, characterized by high levels of plasma triglycerides and LDL-cholesterol, and reduced HDL-cholesterol. It can increase plasma cholesterol levels by promoting hepatic cholesterol synthesis and disturbing lipolytic enzymes, blocking lipid uptake from circulation.
  • Used to induce hyperlipidemia in animals
  • Triggers detachment of HEK293 cells
  • Induces nuclear fragmentation and apoptotic nuclei
  • Increases risk of pulmonary hemorrhage and causes cytotoxicity
  • Reduces AChE and MAO enzyme activities in rat plasma and brain
  • Leads to reduction in plasma urea, creatinine, and bilirubin
  • Appearance: liquid
  • Color: colorless to light yellow
Catalog No. 50-004-14042