Learn More
Medchemexpress LLC Tylvalosin (tartrate) | 63428-13-7 | 96.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128423100MG
Encompass_Preferred
Tylvalosin (Acetylisovaleryltylosin) tartrate is an orally active, broad-spectrum macrolide antibiotic with antimicrobial activity. It is an antiviral agent useful in studying PRRSV infection and induces apoptosis. Tylvalosin tartrate also exhibits anti-inflammatory activity, relieves oxidative stress, and alleviates acute lung injury by inhibiting NF-κB activation.
  • Orally active, broad-spectrum macrolide antibiotic
  • Antiviral agent for PRRSV infection studies
  • Induces apoptosis
  • Anti-inflammatory activity
  • Relieves oxidative stress
  • Alleviates acute lung injury by inhibiting NF-κB activation
Catalog No. 50-003-42062