Learn More
Medchemexpress LLC Tylvalosin (tartrate) | 63428-13-7 | 96.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1284231G
Encompass_Preferred
Tylvalosin (Acetylisovaleryltylosin) tartrate is an orally active, broad-spectrum macrolide antibiotic with antimicrobial activity. It acts as an antiviral agent useful for studying PRRSV infection and induces apoptosis. The compound also demonstrates anti-inflammatory activity, relieves oxidative stress, and alleviates acute lung injury via NF-κB activation inhibition.
  • Orally active, broad-spectrum macrolide antibiotic
  • Exhibits antimicrobial and antiviral activity
  • Useful in studying PRRSV infection and inducing apoptosis
  • Shows anti-inflammatory properties
  • Relieves oxidative stress
  • Alleviates acute lung injury by inhibiting NF-κB activation
Catalog No. 50-003-41202