Learn More
Omega Engineering, Inc. Hermetically Sealed Tip Insulated Thermocouple, Type T, 36in, 24 Gauge
Supplier:  Omega Engineering, Inc. HSTCTTT24S36SMPWCC
Encompass
These PFA insulated thermocouple wire sensors are hermetically sealed at the sensor tip to provide continuous PFA protection over the measurement junction. This hermetic seal design provides a small, flexible, and electrically isolated thermocouple, suited to a variety of applications.
Catalog No. NC1802444