Learn More
Medchemexpress LLC Tyr-Gly-Gly-Phe-Met-OH | 58569-55-4 | 99.6% | 573.66 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP00735MG
Encompass_Preferred
Tyr-Gly-Gly-Phe-Met-OH, also known as Met-Enkephalin, is an endogenous neuropeptide that plays a crucial role in neuroendocrine and immune systems. It regulates human immune function and inhibits tumor growth by binding to the opioid receptor. This product is intended for research use only and not for medical applications.
  • Regulates human immune function
  • Inhibits tumor growth
  • Binds to opioid receptor
  • Endogenous neuropeptide
  • Suitable for research applications
Catalog No. 50-004-26490