Learn More
Medchemexpress LLC Tyr-Gly-Gly-Phe-Met-OH (Met-Enkephalin) | 58569-55-4 | MFCD00076415 | 99.56% | 573.66 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0073
Encompass_Preferred
Tyr-Gly-Gly-Phe-Met-OH, also known as Met-Enkephalin, is an opioid receptor activator that plays a crucial role in both neuroendocrine and immune systems. It is believed to possess immunoregulatory activity and potential cancer biotherapy activity by binding to opioid receptors on immune and cancer cells. This interaction may also alter the tumor microenvironment, revealing a connection between the endocrine and immune systems.
  • Regulates human immune function.
  • Inhibits tumor growth.
  • Binds to the opioid receptor.
  • Endogenous neuropeptide.
  • Agonist activity at human recombinant delta opioid receptor expressed in HEK293 cells.
  • Agonist activity at mu opioid receptor.
Catalog No. 50-004-26969